The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316.3 | NP_001307.2 | 7 - 8 | 865 | 175 |
XM_011528599.2 | XP_011526901.1 | 7 - 8 | 799 | 175 | |
GenBank mRNA | AF053641.1 | - | 7 - 8 | 799 | 175 |
AK297314.1 | - | 6 - 7 | 698 | 175 | |
AK299549.1 | - | 2 - 3 | 170 | 175 | |
AK312306.1 | - | 7 - 8 | 796 | 175 | |
BC108309.1 | - | 7 - 8 | 811 | 175 | |
BC109313.2 | - | 7 - 8 | 778 | 175 | |
BC109314.2 | - | 7 - 8 | 778 | 175 | |
U33286.1 | - | 7 - 8 | 799 | 175 |