The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002915.3 | NP_002906.1 | 5 - 6 | 684 | 150 |
NM_181558.2 | NP_853536.2 | 5 - 6 | 684 | 150 | |
XM_011535172.2 | XP_011533474.1 | 5 - 6 | 704 | 150 | |
XM_011535173.2 | XP_011533475.1 | 5 - 6 | 704 | 150 | |
XM_011535174.2 | XP_011533476.1 | 5 - 6 | 704 | 150 | |
XM_011535175.1 | XP_011533477.1 | 5 - 6 | 704 | 150 | |
XM_017020680.1 | XP_016876169.1 | 5 - 6 | 704 | 150 | |
XM_017020681.1 | XP_016876170.1 | 5 - 6 | 704 | 150 | |
XM_017020682.1 | XP_016876171.1 | 5 - 6 | 704 | 150 | |
XM_017020683.1 | XP_016876172.1 | 5 - 6 | 704 | 150 | |
GenBank mRNA | AK296527.1 | - | 5 - 6 | 625 | 150 |
AK312544.1 | - | 5 - 6 | 601 | 150 | |
BC000149.2 | - | 5 - 6 | 615 | 150 | |
BT019668.1 | - | 5 - 6 | 574 | 150 | |
CX786577.1 | - | 4 - 5 | 426 | 150 | |
L07541.1 | - | 5 - 6 | 583 | 150 |