The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000301.3 | NP_000292.1 | 12 - 13 | 1691 | 82 |
GenBank mRNA | AK298338.1 | - | 8 - 9 | 1047 | 82 |
AK308036.1 | - | 11 - 12 | 1167 | 82 | |
AK312966.1 | - | 12 - 13 | 1642 | 82 | |
BC060513.1 | - | 12 - 13 | 1617 | 82 | |
K02922.1 | - | 5 - 6 | 706 | 82 | |
M74220.1 | - | 12 - 13 | 1628 | 82 | |
X05199.1 | - | 12 - 13 | 1633 | 82 |