The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015278.3 | NP_056093.3 | 3 - 4 | 813 | 68 |
XM_011535640.1 | XP_011533942.1 | 3 - 4 | 256 | 68 | |
XM_017010598.1 | XP_016866087.1 | 3 - 4 | 1045 | 68 | |
XM_017010599.1 | XP_016866088.1 | 3 - 4 | 368 | 68 | |
XM_017010600.1 | XP_016866089.1 | 3 - 4 | 256 | 68 | |
XM_017010601.1 | XP_016866090.1 | 2 - 3 | 470 | 68 | |
XM_017010607.1 | XP_016866096.1 | 3 - 4 | 371 | 68 | |
GenBank mRNA | AB018333.1 | - | 3 - 4 | 556 | 68 |
AJ507735.1 | - | 3 - 4 | 344 | 68 | |
AK025495.1 | - | 3 - 4 | 813 | 68 | |
BC063279.1 | - | 3 - 4 | 282 | 68 |