The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018207.2 | NP_060677.2 | 2 - 3 | 1141 | 72 |
XM_011541705.2 | XP_011540007.1 | 2 - 3 | 761 | 72 | |
XM_017001629.1 | XP_016857118.1 | 3 - 4 | 991 | 72 | |
GenBank mRNA | AK001621.1 | - | 2 - 3 | 737 | 72 |
AK122896.1 | - | 2 - 3 | 1121 | 72 | |
AK300177.1 | - | 2 - 3 | 239 | 72 | |
AX775831.1 | - | 2 - 3 | 545 | 72 | |
BC001222.1 | - | 2 - 3 | 536 | 72 | |
BC007999.1 | - | 2 - 3 | 536 | 72 | |
BC011689.2 | - | 2 - 3 | 536 | 72 | |
BC012152.1 | - | 2 - 3 | 536 | 72 |