Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMC6
Entrez Gene ID
11322
Gene Name
transmembrane channel like 6
Gene Aliases
EV1, EVER1, EVIN1, LAK-4P
Chromosome Location
Chr.17: 78108775 - 78132427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005256995.1 XP_005257052.1 1 - 2 143 149

View Full Product Details