The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172250.2 | NP_758454.1 | 2 - 3 | 521 | 146 |
XM_011531684.2 | XP_011529986.1 | 2 - 3 | 599 | 146 | |
XM_011531685.2 | XP_011529987.1 | 2 - 3 | 642 | 146 | |
GenBank mRNA | AK126662.1 | - | 2 - 3 | 521 | 146 |
BC101178.2 | - | 1 - 2 | 483 | 146 | |
BC101179.2 | - | 1 - 2 | 483 | 146 | |
BC101180.2 | - | 1 - 2 | 483 | 146 | |
BC101181.2 | - | 1 - 2 | 483 | 146 |