Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR36
Entrez Gene ID
134430
Gene Name
WD repeat domain 36
Gene Aliases
GLC1G, TA-WDRP, TAWDRP, UTP21
Chromosome Location
Chr.5: 111092172 - 111130502 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139281.2 NP_644810.1 18 - 19 2283 146
GenBank mRNA AF385437.1 - 18 - 19 2283 146
AK074202.1 - 12 - 13 1333 146
AK095312.1 - 18 - 19 2141 146
AL832494.1 - 17 - 18 2085 146
BC133025.1 - 18 - 19 2243 146
BC136517.1 - 18 - 19 2243 146
CR749211.1 - 13 - 14 1483 146

View Full Product Details