The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199148.1 | NP_001186077.1 | 9 - 10 | 1126 | 134 |
NM_173512.2 | NP_775783.1 | 8 - 9 | 1060 | 134 | |
XM_005246350.4 | XP_005246407.1 | 10 - 11 | 1279 | 134 | |
XM_006712337.3 | XP_006712400.1 | 9 - 10 | 1275 | 134 | |
XM_011510737.2 | XP_011509039.1 | 9 - 10 | 1371 | 134 | |
XM_011510742.2 | XP_011509044.1 | 7 - 8 | 1067 | 134 | |
XM_017003455.1 | XP_016858944.1 | 10 - 11 | 1515 | 134 | |
XM_017003456.1 | XP_016858945.1 | 9 - 10 | 1449 | 134 | |
XM_017003457.1 | XP_016858946.1 | 9 - 10 | 1440 | 134 | |
XM_017003458.1 | XP_016858947.1 | 9 - 10 | 1400 | 134 | |
XM_017003459.1 | XP_016858948.1 | 8 - 9 | 1138 | 134 | |
XM_017003460.1 | XP_016858949.1 | 10 - 11 | 1515 | 134 | |
XM_017003461.1 | XP_016858950.1 | 10 - 11 | 1515 | 134 | |
XM_017003462.1 | XP_016858951.1 | 10 - 11 | 1515 | 134 | |
XM_017003463.1 | XP_016858952.1 | 9 - 10 | 912 | 134 | |
XM_017003464.1 | XP_016858953.1 | 6 - 7 | 1124 | 134 | |
XM_017003465.1 | XP_016858954.1 | 5 - 6 | 652 | 134 | |
GenBank mRNA | AK097141.1 | - | 8 - 9 | 1060 | 134 |
AK293998.1 | - | 9 - 10 | 1087 | 134 | |
BC125157.1 | - | 11 - 12 | 1053 | 134 |