Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SASH1
Entrez Gene ID
23328
Gene Name
SAM and SH3 domain containing 1
Gene Aliases
SH3D6A, dJ323M4, dJ323M4.1
Chromosome Location
Chr.6: 148193440 - 148552049 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011535640.1 XP_011533942.1 1 - 2 73 60
XM_017010600.1 XP_016866089.1 1 - 2 73 60

View Full Product Details