Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CWC27
Entrez Gene ID
10283
Gene Name
CWC27 spliceosome associated protein homolog
Gene Aliases
NY-CO-10, SDCCAG-10, SDCCAG10
Chromosome Location
Chr.5: 64768918 - 65018763 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297644.1 NP_001284573.1 2 - 3 365 100
NM_001297645.1 NP_001284574.1 2 - 3 365 100
NM_001318000.1 NP_001304929.1 2 - 3 365 100
NM_005869.3 NP_005860.2 2 - 3 365 100
XM_011543095.2 XP_011541397.2 2 - 3 334 100
XM_011543096.1 XP_011541398.1 2 - 3 334 100
XM_017008943.1 XP_016864432.1 2 - 3 334 100
GenBank mRNA AF039692.1 - 2 - 3 317 101
AK225105.1 - 2 - 3 332 100
AK225161.1 - 2 - 3 337 100
AK308223.1 - 2 - 3 334 100
AK312954.1 - 2 - 3 334 100
AY358569.1 - 2 - 3 326 100
BC012117.1 - 2 - 3 318 100
BX647971.1 - 2 - 3 366 100

View Full Product Details