The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032286.2 | NP_115662.2 | 1 - 2 | 167 | 102 |
GenBank mRNA | AY277599.1 | - | 1 - 2 | 124 | 102 |
AY453397.1 | - | 1 - 2 | 165 | 102 | |
AY598325.1 | - | 1 - 2 | 124 | 102 | |
BC003353.1 | - | 1 - 2 | 167 | 102 |