The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004073.3 | NP_004064.2 | 4 - 5 | 803 | 68 |
GenBank mRNA | AB209875.1 | - | 5 - 6 | 490 | 68 |
AJ293866.1 | - | 4 - 5 | 803 | 68 | |
BC013899.2 | - | 4 - 5 | 664 | 68 | |
U56998.1 | - | 4 - 5 | 483 | 68 |