Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCND1
Entrez Gene ID
3750
Gene Name
potassium voltage-gated channel subfamily D member 1
Gene Aliases
KV4.1
Chromosome Location
Chr.X: 48960983 - 48972099 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004979.5 NP_004970.3 3 - 4 2942 86
XM_011543910.2 XP_011542212.1 4 - 5 1850 86
XM_017029508.1 XP_016884997.1 4 - 5 1681 86
XM_017029509.1 XP_016884998.1 4 - 5 1678 86
GenBank mRNA AB021865.1 - 3 - 4 2667 86
AF166003.1 - 3 - 4 1419 86
AJ005898.1 - 3 - 4 1369 86
AK094431.1 - 2 - 3 1528 86
AK315092.1 - 3 - 4 1853 86
BC045659.1 - 3 - 4 2940 86

View Full Product Details