Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GIPR
Entrez Gene ID
2696
Gene Name
gastric inhibitory polypeptide receptor
Gene Aliases
PGQTL2
Chromosome Location
Chr.19: 45668187 - 45683724 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000164.3 NP_000155.1 6 - 7 586 78
NM_001308418.1 NP_001295347.1 5 - 6 478 78
XM_011526709.2 XP_011525011.1 6 - 7 643 78
XM_011526710.2 XP_011525012.1 7 - 8 724 78
XM_011526711.2 XP_011525013.1 5 - 6 535 78
XM_011526714.2 XP_011525016.1 3 - 4 319 78
XM_011526715.2 XP_011525017.1 3 - 4 329 78
XM_011526716.2 XP_011525018.1 6 - 7 643 78
GenBank mRNA BC093723.1 - 6 - 7 573 78
BC095494.1 - 5 - 6 485 78
BC101673.1 - 6 - 7 573 78
BC143300.1 - 5 - 6 466 78
BC143302.1 - 6 - 7 573 78
S79852.1 - 6 - 7 581 78
U39231.1 - 6 - 7 586 78

View Full Product Details