Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP6V1E1
Entrez Gene ID
529
Gene Name
ATPase H+ transporting V1 subunit E1
Gene Aliases
ATP6E, ATP6E2, ATP6V1E, P31, Vma4
Chromosome Location
Chr.22: 17592136 - 17628822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039366.1 NP_001034455.1 3 - 4 393 86
NM_001696.3 NP_001687.1 4 - 5 459 86
GenBank mRNA AK294623.1 - 3 - 4 320 86
AK312394.1 - 4 - 5 380 86
BC004443.1 - 4 - 5 361 86
BI546032.1 - 4 - 5 411 86
BQ888820.1 - 3 - 4 338 86
BT007128.1 - 4 - 5 272 86
CK005516.1 - 4 - 5 462 86
CR456385.1 - 4 - 5 325 86
X71491.1 - 4 - 5 328 86
X76228.1 - 4 - 5 347 86

View Full Product Details