The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039366.1 | NP_001034455.1 | 6 - 7 | 650 | 102 |
NM_001039367.1 | NP_001034456.1 | 6 - 7 | 626 | 102 | |
NM_001696.3 | NP_001687.1 | 7 - 8 | 716 | 102 | |
GenBank mRNA | AK294623.1 | - | 6 - 7 | 577 | 102 |
AK312394.1 | - | 7 - 8 | 637 | 102 | |
AK315941.1 | - | 6 - 7 | 552 | 102 | |
BC004443.1 | - | 7 - 8 | 618 | 102 | |
BE735148.1 | - | 6 - 7 | 572 | 102 | |
BQ888820.1 | - | 6 - 7 | 595 | 102 | |
BT007128.1 | - | 7 - 8 | 529 | 102 | |
CR456385.1 | - | 7 - 8 | 582 | 102 | |
X71491.1 | - | 7 - 8 | 585 | 102 | |
X76228.1 | - | 7 - 8 | 604 | 102 |