Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR18
Entrez Gene ID
57418
Gene Name
WD repeat domain 18
Gene Aliases
Ipi3, R32184_1
Chromosome Location
Chr.19: 982980 - 994570 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024100.3 NP_077005.2 9 - 10 1195 78
XM_017027027.1 XP_016882516.1 11 - 12 1721 78
GenBank mRNA AK131009.1 - 3 - 4 338 78
BC000040.2 - 9 - 10 1180 78
BC001648.2 - 9 - 10 1175 78

View Full Product Details