Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC11
Entrez Gene ID
55735
Gene Name
DnaJ heat shock protein family (Hsp40) member C11
Gene Aliases
dJ126A5.1
Chromosome Location
Chr.1: 6634168 - 6701906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018198.3 NP_060668.2 10 - 11 1206 90
XM_017001745.1 XP_016857234.1 9 - 10 1059 90
GenBank mRNA AF306695.1 - 10 - 11 1120 90
AK001599.1 - 10 - 11 1115 90
AK095386.1 - 2 - 3 137 90
AK294539.1 - 9 - 10 1029 90
AK298378.1 - 10 - 11 1092 90
AL833841.1 - 10 - 11 1080 90
AY336749.1 - 10 - 11 1116 90
BC006086.1 - 10 - 11 1102 90
BC008772.1 - 10 - 11 1107 90

View Full Product Details