Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABHD1
Entrez Gene ID
84696
Gene Name
abhydrolase domain containing 1
Gene Aliases
LABH1
Chromosome Location
Chr.2: 27123780 - 27130812 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032604.3 NP_115993.3 5 - 6 781 102
XM_011533135.2 XP_011531437.1 5 - 6 747 102
XM_011533136.2 XP_011531438.1 5 - 6 747 102
XM_011533137.2 XP_011531439.1 5 - 6 747 102
XM_011533138.2 XP_011531440.1 4 - 5 629 102
GenBank mRNA AK093447.1 - 5 - 6 747 102
AY033290.1 - 5 - 6 654 102
BC039576.1 - 5 - 6 735 102

View Full Product Details