Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XPO7
Entrez Gene ID
23039
Gene Name
exportin 7
Gene Aliases
EXP7, RANBP16
Chromosome Location
Chr.8: 21919669 - 22006585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015024.4 NP_055839.3 12 - 13 1575 77
XM_017013244.1 XP_016868733.1 12 - 13 1829 77
XM_017013245.1 XP_016868734.1 12 - 13 1596 77
XM_017013246.1 XP_016868735.1 12 - 13 1798 77
XM_017013247.1 XP_016868736.1 12 - 13 1989 77
GenBank mRNA AB018288.1 - 9 - 10 939 77
AF064729.1 - 12 - 13 1485 77
AK291472.1 - 12 - 13 1490 77
AK299848.1 - 8 - 9 1011 77
BC030785.1 - 12 - 13 1574 77

View Full Product Details