The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015024.4 | NP_055839.3 | 22 - 23 | 2534 | 67 |
XM_017013244.1 | XP_016868733.1 | 22 - 23 | 2788 | 67 | |
XM_017013245.1 | XP_016868734.1 | 22 - 23 | 2555 | 67 | |
XM_017013246.1 | XP_016868735.1 | 22 - 23 | 2757 | 67 | |
GenBank mRNA | AB018288.1 | - | 19 - 20 | 1898 | 67 |
AF064729.1 | - | 22 - 23 | 2444 | 67 | |
AK291472.1 | - | 22 - 23 | 2449 | 67 | |
AK299848.1 | - | 18 - 19 | 1970 | 67 | |
BC030785.1 | - | 22 - 23 | 2533 | 67 |