Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XPO7
Entrez Gene ID
23039
Gene Name
exportin 7
Gene Aliases
EXP7, RANBP16
Chromosome Location
Chr.8: 21919669 - 22006585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015024.4 NP_055839.3 26 - 27 3137 76
XM_017013244.1 XP_016868733.1 26 - 27 3391 76
XM_017013245.1 XP_016868734.1 26 - 27 3158 76
XM_017013246.1 XP_016868735.1 26 - 27 3360 76
GenBank mRNA AB018288.1 - 23 - 24 2501 76
AF064729.1 - 26 - 27 3047 76
AK291472.1 - 26 - 27 3052 76
AK299848.1 - 22 - 23 2573 76
BC014219.2 - 3 - 4 396 76
BC030785.1 - 26 - 27 3136 76

View Full Product Details