Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RER1
Entrez Gene ID
11079
Gene Name
retention in endoplasmic reticulum sorting receptor 1
Gene Aliases
-
Chromosome Location
Chr.1: 2391314 - 2405444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007033.4 NP_008964.3 6 - 7 694 92
XM_005244713.4 XP_005244770.2 6 - 7 1515 92
XM_006710306.2 XP_006710369.1 7 - 8 776 92
XM_011540542.1 XP_011538844.1 7 - 8 714 92
XM_011540543.1 XP_011538845.1 6 - 7 562 92
XM_017000131.1 XP_016855620.1 6 - 7 2045 92
XM_017000132.1 XP_016855621.1 7 - 8 804 92
XM_017000133.1 XP_016855622.1 7 - 8 887 92
XM_017000134.1 XP_016855623.1 8 - 9 801 92
XM_017000135.1 XP_016855624.1 6 - 7 801 92
GenBank mRNA AF091071.1 - 6 - 7 688 92
AF157324.1 - 6 - 7 633 92
AJ001421.1 - 5 - 6 503 92
AK095750.1 - 5 - 6 2289 92
AK289658.1 - 6 - 7 628 92
BC004965.1 - 6 - 7 629 92
BT007224.1 - 5 - 6 503 92
CR456806.1 - 5 - 6 503 92

View Full Product Details