The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003920.3 | NP_003911.2 | 5 - 6 | 599 | 64 |
XM_011538939.2 | XP_011537241.1 | 5 - 6 | 577 | 64 | |
GenBank mRNA | AB015597.1 | - | 4 - 5 | 431 | 64 |
AF098162.1 | - | 5 - 6 | 573 | 64 | |
AK296869.1 | - | 5 - 6 | 579 | 64 | |
BC031514.1 | - | 5 - 6 | 562 | 64 | |
BC039842.1 | - | 5 - 6 | 586 | 64 | |
BC050557.1 | - | 5 - 6 | 626 | 64 |