The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003920.3 | NP_003911.2 | 6 - 7 | 696 | 84 |
GenBank mRNA | AB015597.1 | - | 5 - 6 | 528 | 84 |
AF098162.1 | - | 6 - 7 | 670 | 84 | |
AK296869.1 | - | 6 - 7 | 676 | 84 | |
BC031514.1 | - | 6 - 7 | 659 | 84 | |
BC050557.1 | - | 6 - 7 | 723 | 84 |