Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THOP1
Entrez Gene ID
7064
Gene Name
thimet oligopeptidase 1
Gene Aliases
EP24.15, MEPD_HUMAN, MP78, TOP
Chromosome Location
Chr.19: 2785460 - 2813604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003249.4 NP_003240.1 11 - 12 1971 64
XM_011528228.2 XP_011526530.1 8 - 9 2918 64
GenBank mRNA AK093392.1 - 2 - 3 1089 64
AK300552.1 - 8 - 9 1453 64
BC000135.2 - 11 - 12 1897 64
BC000583.2 - 9 - 10 1476 64
BC002391.2 - 11 - 12 1923 64
BC013878.2 - 11 - 12 1887 64
U29366.1 - 11 - 12 1855 64
Z50115.1 - 11 - 12 1895 64

View Full Product Details