The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000337.5 | NP_000328.2 | 1 - 2 | 474 | 58 |
NM_172244.2 | NP_758447.1 | 1 - 2 | 474 | 58 | |
XM_005265967.2 | XP_005266024.1 | 1 - 2 | 66 | 58 | |
GenBank mRNA | AK292794.1 | - | 1 - 2 | 66 | 58 |
BC020740.1 | - | 1 - 2 | 67 | 58 | |
BQ434817.1 | - | 1 - 2 | 474 | 58 |