Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GJA10
Entrez Gene ID
84694
Gene Name
gap junction protein alpha 10
Gene Aliases
CX62
Chromosome Location
Chr.6: 89894469 - 89896100 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032602.1 NP_115991.1 1 - 1 1034 90
GenBank mRNA AF296766.1 - 1 - 1 1034 90

View Full Product Details