The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000553.4 | NP_000544.2 | 29 - 30 | 4246 | 68 |
XM_011544639.2 | XP_011542941.1 | 28 - 29 | 3647 | 68 | |
XM_011544640.1 | XP_011542942.1 | 17 - 18 | 1959 | 68 | |
GenBank mRNA | AB209652.1 | - | 15 - 16 | 1689 | 68 |
AF091214.1 | - | 29 - 30 | 3689 | 68 | |
AL833572.1 | - | 17 - 18 | 2092 | 68 | |
AY818673.1 | - | 28 - 29 | 3458 | 68 |