The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000280.4 | NP_000271.1 | 4 - 5 | 568 | 55 |
NM_001127612.1 | NP_001121084.1 | 4 - 5 | 482 | 55 | |
NM_001258462.1 | NP_001245391.1 | 4 - 5 | 482 | 55 | |
NM_001258463.1 | NP_001245392.1 | 4 - 5 | 420 | 55 | |
NM_001258464.1 | NP_001245393.1 | 4 - 5 | 470 | 55 | |
NM_001258465.1 | NP_001245394.1 | 3 - 4 | 456 | 55 | |
NM_001310158.1 | NP_001297087.1 | 4 - 5 | 523 | 55 | |
NM_001310159.1 | NP_001297088.1 | 2 - 3 | 141 | 55 | |
NM_001604.5 | NP_001595.2 | 4 - 5 | 470 | 55 | |
GenBank mRNA | AB593092.1 | - | 4 - 5 | 421 | 55 |
AB593093.1 | - | 4 - 5 | 420 | 55 | |
AB593094.1 | - | 4 - 5 | 523 | 55 | |
AK074881.1 | - | 3 - 4 | 903 | 55 | |
AK314470.1 | - | 4 - 5 | 460 | 55 | |
AY047583.1 | - | 2 - 3 | 143 | 55 | |
BC011953.1 | - | 4 - 5 | 354 | 55 | |
BX440968.2 | - | 4 - 5 | 890 | 55 | |
BX640762.1 | - | 4 - 5 | 449 | 55 | |
DA079367.1 | - | 3 - 4 | 456 | 55 | |
DA141443.1 | - | 4 - 5 | 388 | 55 | |
DA183294.1 | - | 4 - 5 | 482 | 55 | |
GQ141695.1 | - | 1 - 2 | 61 | 55 | |
M77844.1 | - | 3 - 4 | 375 | 55 | |
M93650.1 | - | 4 - 5 | 430 | 55 |