Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARD8
Entrez Gene ID
22900
Gene Name
caspase recruitment domain family member 8
Gene Aliases
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN
Chromosome Location
Chr.19: 48203148 - 48256269 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184900.1 NP_001171829.1 10 - 11 1396 54
NM_001184901.1 NP_001171830.1 10 - 11 1343 54
NM_001184902.1 NP_001171831.1 12 - 13 1617 54
NM_001184903.1 NP_001171832.1 10 - 11 1351 54
NM_014959.3 NP_055774.2 11 - 12 1454 54
NR_033678.1 9 - 10 1313 54
NR_033679.1 9 - 10 1355 54
NR_033680.1 8 - 9 1171 54
XM_006723091.3 XP_006723154.1 12 - 13 1607 54
XM_006723092.3 XP_006723155.1 14 - 15 2145 54
XM_006723093.3 XP_006723156.1 12 - 13 1928 54
XM_006723094.3 XP_006723157.1 12 - 13 1587 54
XM_006723095.3 XP_006723158.1 12 - 13 1574 54
XM_006723096.3 XP_006723159.1 13 - 14 1996 54
XM_006723097.3 XP_006723160.1 11 - 12 1779 54
XM_006723098.3 XP_006723161.1 11 - 12 1566 54
XM_006723099.3 XP_006723162.1 10 - 11 1452 54
XM_006723101.2 XP_006723164.1 11 - 12 1469 54
XM_006723102.3 XP_006723165.1 10 - 11 1409 54
XM_006723104.3 XP_006723167.1 11 - 12 1466 54
XM_006723106.3 XP_006723169.1 11 - 12 1797 54
XM_006723107.3 XP_006723170.1 10 - 11 1322 54
XM_006723109.3 XP_006723172.1 10 - 11 1324 54
XM_011526641.2 XP_011524943.1 15 - 16 2452 54
XM_011526642.2 XP_011524944.1 14 - 15 2347 54
XM_011526643.2 XP_011524945.1 13 - 14 2034 54
XM_011526644.2 XP_011524946.1 11 - 12 1619 54
XM_011526646.2 XP_011524948.1 12 - 13 2093 54
XM_011526647.2 XP_011524949.1 11 - 12 1982 54
XM_011526648.2 XP_011524950.1 12 - 13 2099 54
XM_011526653.2 XP_011524955.1 11 - 12 1987 54
XM_017026482.1 XP_016881971.1 12 - 13 1932 54
XM_017026483.1 XP_016881972.1 12 - 13 1885 54
XM_017026484.1 XP_016881973.1 12 - 13 1893 54
XM_017026486.1 XP_016881975.1 10 - 11 1449 54
XM_017026487.1 XP_016881976.1 9 - 10 1818 54
XM_017026488.1 XP_016881977.1 13 - 14 2034 54
XM_017026489.1 XP_016881978.1 12 - 13 1619 54
XM_017026492.1 XP_016881981.1 11 - 12 1759 54
XM_017026493.1 XP_016881982.1 10 - 11 1651 54
XM_017026494.1 XP_016881983.1 11 - 12 2265 54
XM_017026495.1 XP_016881984.1 10 - 11 1407 54
XM_017026496.1 XP_016881985.1 10 - 11 1327 54
XM_017026497.1 XP_016881986.1 9 - 10 1267 54
XM_017026498.1 XP_016881987.1 9 - 10 1264 54
XM_017026499.1 XP_016881988.1 8 - 9 3037 54
GenBank mRNA AB023172.1 - 9 - 10 1348 54
AF322184.1 - 9 - 10 1348 54
AF331519.1 - 7 - 8 1035 54
AF405558.1 - 7 - 8 1035 54
AF511652.1 - 8 - 9 1206 54
AK297045.1 - 10 - 11 1337 54
AK297069.1 - 10 - 11 1333 54
AY026322.1 - 7 - 8 1035 54
BX641091.1 - 4 - 5 998 54
BX648568.1 - 11 - 12 3500 54
EU118120.1 - 10 - 11 1396 54
EU118121.1 - 9 - 10 1355 54
EU118122.1 - 7 - 8 1196 54

View Full Product Details