The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003801.1 | NP_001003801.1 | 12 - 13 | 1498 | 84 |
NM_001003802.1 | NP_001003802.1 | 13 - 14 | 1487 | 84 | |
NM_003078.3 | NP_003069.2 | 13 - 14 | 1526 | 84 | |
XM_011516521.2 | XP_011514823.1 | 12 - 13 | 1605 | 84 | |
XM_017012556.1 | XP_016868045.1 | 12 - 13 | 1341 | 84 | |
GenBank mRNA | AK127595.1 | - | 13 - 14 | 2034 | 84 |
AY450430.1 | - | 12 - 13 | 1802 | 84 | |
AY450431.1 | - | 13 - 14 | 1487 | 84 | |
BC002628.2 | - | 13 - 14 | 1487 | 84 | |
BC110350.1 | - | 12 - 13 | 1417 | 84 | |
BX648385.1 | - | 11 - 12 | 2249 | 84 | |
U66619.1 | - | 13 - 14 | 1535 | 84 |