The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308440.1 | NP_001295369.1 | 6 - 7 | 871 | 93 |
NM_020828.1 | NP_065879.1 | 6 - 7 | 871 | 93 | |
XM_011526462.2 | XP_011524764.1 | 6 - 7 | 867 | 93 | |
XM_011526463.2 | XP_011524765.2 | 6 - 7 | 1600 | 93 | |
GenBank mRNA | AB037852.1 | - | 6 - 7 | 871 | 93 |
AF507045.1 | - | 6 - 7 | 871 | 93 | |
AK289583.1 | - | 6 - 7 | 856 | 93 | |
BC028416.2 | - | 6 - 7 | 866 | 93 | |
BC099903.1 | - | 6 - 7 | 866 | 93 | |
BC127002.1 | - | 6 - 7 | 856 | 93 | |
BC127003.1 | - | 6 - 7 | 856 | 93 |