The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184993.1 | NP_001171922.1 | 3 - 4 | 351 | 118 |
NM_031277.2 | NP_112567.2 | 3 - 4 | 351 | 118 | |
XM_006719846.3 | XP_006719909.1 | 3 - 4 | 645 | 118 | |
XM_006719849.2 | XP_006719912.1 | 3 - 4 | 362 | 118 | |
XM_011535152.2 | XP_011533454.1 | 7 - 8 | 2270 | 118 | |
XM_011535155.2 | XP_011533457.1 | 6 - 7 | 1006 | 118 | |
XM_011535156.2 | XP_011533458.1 | 7 - 8 | 2270 | 118 | |
XM_011535157.2 | XP_011533459.1 | 7 - 8 | 2270 | 118 | |
XM_011535158.2 | XP_011533460.1 | 7 - 8 | 2270 | 118 | |
XM_011535159.2 | XP_011533461.1 | 7 - 8 | 2270 | 118 | |
XM_011535160.2 | XP_011533462.1 | 7 - 8 | 2270 | 118 | |
XM_017020676.1 | XP_016876165.1 | 3 - 4 | 314 | 118 | |
GenBank mRNA | AF285602.1 | - | 3 - 4 | 315 | 118 |
AF285603.1 | - | 3 - 4 | 315 | 118 | |
AK302435.1 | - | 3 - 4 | 327 | 118 | |
BC146822.1 | - | 3 - 4 | 343 | 118 |