The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175056.1 | NP_778226.1 | 5 - 6 | 721 | 119 |
XM_017005703.1 | XP_016861192.1 | 6 - 7 | 903 | 119 | |
XM_017005704.1 | XP_016861193.1 | 5 - 6 | 759 | 119 | |
GenBank mRNA | AY090780.1 | - | 5 - 6 | 721 | 119 |
BC031261.1 | - | 12 - 13 | 1455 | 119 | |
BC143817.1 | - | 6 - 7 | 817 | 119 |