Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH6
Entrez Gene ID
1768
Gene Name
dynein axonemal heavy chain 6
Gene Aliases
DNHL1, Dnahc6, HL-2, HL2
Chromosome Location
Chr.2: 84459588 - 84819589 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001370.1 NP_001361.1 13 - 14 2214 126
XM_006711956.2 XP_006712019.1 13 - 14 2319 126
XM_011532649.2 XP_011530951.1 13 - 14 2168 126
XM_011532650.2 XP_011530952.1 13 - 14 2169 126
XM_011532652.1 XP_011530954.1 13 - 14 2214 126
XM_011532653.1 XP_011530955.1 13 - 14 2214 126
XM_011532654.1 XP_011530956.1 12 - 13 1947 126
XM_011532655.2 XP_011530957.1 9 - 10 1236 126
XM_011532657.1 XP_011530959.1 7 - 8 975 126
XM_011532660.1 XP_011530962.1 13 - 14 2214 126
XM_017003521.1 XP_016859010.1 13 - 14 2214 126
XM_017003522.1 XP_016859011.1 8 - 9 2040 126
XM_017003523.1 XP_016859012.1 8 - 9 1207 126
XM_017003524.1 XP_016859013.1 13 - 14 2214 126
GenBank mRNA AK094676.1 - 9 - 10 1427 126
BC104884.1 - 7 - 8 1120 126
BC113424.1 - 7 - 8 1120 126
BC143666.1 - 7 - 8 1120 126

View Full Product Details