Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMTC3
Entrez Gene ID
160418
Gene Name
transmembrane and tetratricopeptide repeat containing 3
Gene Aliases
SMILE
Chromosome Location
Chr.12: 88142296 - 88199887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181783.3 NP_861448.2 12 - 13 1929 103
XM_011537980.2 XP_011536282.1 11 - 12 1704 103
XM_011537981.2 XP_011536283.1 5 - 6 593 103
GenBank mRNA AJ697717.1 - 12 - 13 1799 103
AK074973.1 - 11 - 12 1556 103
AK290436.1 - 12 - 13 1929 103
AK302755.1 - 12 - 13 1929 103
BC117175.1 - 11 - 12 1750 103
BC117177.1 - 11 - 12 1751 103
BX538169.1 - 12 - 13 1902 103
CR749244.1 - 12 - 13 1881 103
CR749309.1 - 12 - 13 1915 103
CR933632.1 - 12 - 13 1880 103
CR936640.1 - 2 - 3 257 103

View Full Product Details