Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMTC3
Entrez Gene ID
160418
Gene Name
transmembrane and tetratricopeptide repeat containing 3
Gene Aliases
SMILE
Chromosome Location
Chr.12: 88142296 - 88199887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181783.3 NP_861448.2 6 - 7 1011 141
XM_011537980.2 XP_011536282.1 5 - 6 786 141
GenBank mRNA AJ697717.1 - 6 - 7 881 141
AK074973.1 - 5 - 6 638 141
AK290436.1 - 6 - 7 1011 141
AK302755.1 - 6 - 7 1011 141
AK310375.1 - 6 - 7 1011 141
AK310393.1 - 6 - 7 1011 141
BC117175.1 - 5 - 6 832 141
BC117177.1 - 5 - 6 833 141
BX538169.1 - 6 - 7 984 141
CR749244.1 - 6 - 7 963 141
CR749309.1 - 6 - 7 997 141
CR933632.1 - 6 - 7 962 141

View Full Product Details