The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017721.4 | NP_060191.3 | 3 - 4 | 609 | 59 |
XM_005259972.2 | XP_005260029.1 | 3 - 4 | 557 | 59 | |
XM_005259973.2 | XP_005260030.1 | 3 - 4 | 557 | 59 | |
XM_005259974.2 | XP_005260031.1 | 3 - 4 | 557 | 59 | |
XM_005259975.2 | XP_005260032.1 | 3 - 4 | 557 | 59 | |
GenBank mRNA | AB097002.1 | - | 3 - 4 | 551 | 59 |
AF536205.1 | - | 5 - 6 | 662 | 59 | |
AK000248.1 | - | - | 129 | 59 | |
AK315068.1 | - | 3 - 4 | 527 | 59 | |
BC064981.1 | - | 3 - 4 | 513 | 59 | |
BC113968.1 | - | 2 - 3 | 238 | 59 |