The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316327.1 | NP_001303256.1 | 14 - 15 | 1540 | 122 |
NM_006254.3 | NP_006245.2 | 14 - 15 | 1707 | 122 | |
NM_212539.1 | NP_997704.1 | 13 - 14 | 1595 | 122 | |
XM_006713257.3 | XP_006713320.1 | 13 - 14 | 1514 | 122 | |
XM_006713259.2 | XP_006713322.1 | 15 - 16 | 1811 | 122 | |
XM_017006855.1 | XP_016862344.1 | 13 - 14 | 1392 | 122 | |
XM_017006856.1 | XP_016862345.1 | 13 - 14 | 1966 | 122 | |
GenBank mRNA | AK130150.1 | - | 9 - 10 | 897 | 122 |
AK294272.1 | - | 11 - 12 | 1166 | 122 | |
AK313216.1 | - | 14 - 15 | 1540 | 122 | |
BC043350.1 | - | 14 - 15 | 1726 | 122 | |
D10495.1 | - | 11 - 12 | 1213 | 122 | |
L07860.1 | - | 13 - 14 | 1412 | 122 | |
L07861.1 | - | 13 - 14 | 1412 | 122 |