The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005914.2 | NP_001005914.1 | 17 - 18 | 2129 | 141 |
NM_001290060.1 | NP_001276989.1 | 16 - 17 | 2086 | 141 | |
NM_001290061.1 | NP_001276990.1 | 16 - 17 | 1935 | 141 | |
NM_001290062.1 | NP_001276991.1 | 10 - 11 | 1352 | 141 | |
NM_001290063.1 | NP_001276992.1 | 10 - 11 | 1226 | 141 | |
NM_004636.3 | NP_004627.1 | 17 - 18 | 2132 | 141 | |
NR_110697.1 | 7 - 8 | 853 | 141 | ||
GenBank mRNA | AB083186.1 | - | 16 - 17 | 1882 | 141 |
AB209322.1 | - | 15 - 16 | 2312 | 141 | |
AF217991.1 | - | 16 - 17 | 1895 | 141 | |
AK092182.1 | - | 7 - 8 | 815 | 141 | |
AK293678.1 | - | 10 - 11 | 1185 | 141 | |
BC009113.1 | - | 13 - 14 | 1836 | 141 | |
BC013975.2 | - | 16 - 17 | 1909 | 141 | |
BC024220.2 | - | 16 - 17 | 1909 | 141 | |
U28369.1 | - | 16 - 17 | 2075 | 141 |