The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031231.3 | NP_112508.3 | 8 - 9 | 826 | 78 |
XM_011528991.1 | XP_011527293.1 | 4 - 5 | 452 | 78 | |
XM_017028015.1 | XP_016883504.1 | 4 - 5 | 451 | 78 | |
XM_017028016.1 | XP_016883505.1 | 4 - 5 | 380 | 78 | |
GenBank mRNA | AF409141.1 | - | 8 - 9 | 774 | 78 |
AK023706.1 | - | 7 - 8 | 1032 | 78 | |
AK124388.1 | - | 4 - 5 | 900 | 78 | |
AK291895.1 | - | 8 - 9 | 756 | 78 | |
BC047673.1 | - | 8 - 9 | 781 | 78 |