Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP2B2
Entrez Gene ID
491
Gene Name
ATPase plasma membrane Ca2+ transporting 2
Gene Aliases
PMCA2, PMCA2a, PMCA2i
Chromosome Location
Chr.3: 10324023 - 10707962 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001331.2 NP_001001331.1 16 - 17 2845 63
NM_001683.3 NP_001674.2 13 - 14 2710 63
XM_005265179.4 XP_005265236.1 18 - 19 3029 63
XM_006713175.3 XP_006713238.1 18 - 19 2913 63
XM_011533752.2 XP_011532054.1 16 - 17 2818 63
XM_017006481.1 XP_016861970.1 19 - 20 3156 63
XM_017006482.1 XP_016861971.1 16 - 17 2847 63
XM_017006483.1 XP_016861972.1 14 - 15 2857 63
XM_017006484.1 XP_016861973.1 16 - 17 2820 63
XM_017006485.1 XP_016861974.1 16 - 17 2952 63
XM_017006486.1 XP_016861975.1 16 - 17 2952 63
XM_017006487.1 XP_016861976.1 15 - 16 2777 63
XM_017006488.1 XP_016861977.1 14 - 15 2859 63
XM_017006489.1 XP_016861978.1 14 - 15 2726 63
XM_017006490.1 XP_016861979.1 13 - 14 2699 63
XM_017006491.1 XP_016861980.1 13 - 14 2816 63
XM_017006492.1 XP_016861981.1 16 - 17 2951 63
GenBank mRNA AB210008.1 - 13 - 14 2783 63
DQ201777.1 - 5 - 6 879 63
L00620.1 - 12 - 13 2289 63
L20977.1 - 15 - 16 2847 63
M97260.1 - 13 - 14 2691 63
S95758.1 - 12 - 13 1974 63
X63575.1 - 15 - 16 2476 63

View Full Product Details