The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025181.3 | NP_079457.2 | 9 - 10 | 1332 | 102 |
NR_104470.1 | 9 - 10 | 1717 | 102 | ||
XM_005263799.3 | XP_005263856.1 | 9 - 10 | 1717 | 102 | |
XM_017005028.1 | XP_016860517.1 | 9 - 10 | 1129 | 102 | |
GenBank mRNA | AF529364.1 | - | 9 - 10 | 918 | 102 |
AK024666.1 | - | 4 - 5 | 459 | 102 | |
AK315719.1 | - | 9 - 10 | 1130 | 102 | |
AY358620.1 | - | 9 - 10 | 1075 | 102 | |
BC018537.1 | - | 9 - 10 | 1342 | 102 |