The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321173.1 | NP_001308102.1 | 5 - 6 | 675 | 83 |
NM_001321174.1 | NP_001308103.1 | 3 - 4 | 511 | 83 | |
NM_001321175.1 | NP_001308104.1 | 6 - 7 | 683 | 83 | |
NM_001321176.1 | NP_001308105.1 | 6 - 7 | 678 | 83 | |
NM_005486.2 | NP_005477.2 | 5 - 6 | 675 | 83 | |
XM_017024002.1 | XP_016879491.1 | 6 - 7 | 595 | 83 | |
GenBank mRNA | AJ010071.1 | - | 5 - 6 | 530 | 83 |
AK223125.1 | - | 5 - 6 | 509 | 83 | |
AK225351.1 | - | 5 - 6 | 527 | 83 | |
AK303912.1 | - | 4 - 5 | 429 | 83 | |
AK303913.1 | - | 5 - 6 | 492 | 83 | |
AK309164.1 | - | 3 - 4 | 370 | 83 | |
AK315039.1 | - | 5 - 6 | 513 | 83 | |
AK315907.1 | - | 3 - 4 | 349 | 83 | |
BC029396.1 | - | 5 - 6 | 509 | 83 |