The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282721.1 | NP_001269650.1 | 2 - 3 | 240 | 116 |
NM_001282723.1 | NP_001269652.1 | 3 - 4 | 396 | 116 | |
NM_021925.3 | NP_068744.1 | 3 - 4 | 396 | 116 | |
NR_104233.1 | 2 - 3 | 240 | 116 | ||
XM_011533014.2 | XP_011531316.1 | 3 - 4 | 423 | 116 | |
GenBank mRNA | AK025473.1 | - | 3 - 4 | 334 | 116 |
AK299244.1 | - | 3 - 4 | 337 | 116 | |
AK300494.1 | - | 2 - 3 | 192 | 116 | |
AK316162.1 | - | 2 - 3 | 199 | 116 | |
BC017473.1 | - | 3 - 4 | 353 | 116 |