Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EBF1
Entrez Gene ID
1879
Gene Name
early B-cell factor 1
Gene Aliases
COE1, EBF, O/E-1, OLF1
Chromosome Location
Chr.5: 158695915 - 159099762 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290360.2 NP_001277289.1 11 - 12 1406 85
NM_001324101.1 NP_001311030.1 11 - 12 1406 85
NM_001324103.1 NP_001311032.1 11 - 12 1403 85
NM_001324106.1 NP_001311035.1 11 - 12 1379 85
NM_001324107.1 NP_001311036.1 11 - 12 1382 85
NM_001324108.1 NP_001311037.1 11 - 12 1379 85
NM_001324109.1 NP_001311038.1 11 - 12 1406 85
NM_001324111.1 NP_001311040.1 11 - 12 1501 85
NM_024007.4 NP_076870.1 11 - 12 1403 85
NM_182708.2 NP_874367.1 10 - 11 1310 85
XM_017009192.1 XP_016864681.1 11 - 12 1406 85
XM_017009193.1 XP_016864682.1 11 - 12 1391 85
XM_017009194.1 XP_016864683.1 11 - 12 1382 85
XM_017009195.1 XP_016864684.1 11 - 12 1379 85
XM_017009196.1 XP_016864685.1 11 - 12 2341 85
XM_017009197.1 XP_016864686.1 11 - 12 1844 85
XM_017009198.1 XP_016864687.1 11 - 12 1763 85
XM_017009199.1 XP_016864688.1 6 - 7 1757 85
XM_017009200.1 XP_016864689.1 6 - 7 18893 85
XM_017009201.1 XP_016864690.1 6 - 7 18890 85
XM_017009204.1 XP_016864693.1 3 - 4 1283 85
GenBank mRNA AK289712.1 - 11 - 12 1390 85
AK303698.1 - 9 - 10 1032 85
AK304438.1 - 11 - 12 1109 85
BC038805.1 - 11 - 12 1414 85
BC041178.1 - 10 - 11 1237 85

View Full Product Details