Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRKX
Entrez Gene ID
5613
Gene Name
protein kinase, X-linked
Gene Aliases
PKX1
Chromosome Location
Chr.X: 3604343 - 3713634 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005044.4 NP_005035.1 5 - 6 1191 72
GenBank mRNA BC041073.1 - 5 - 6 994 72
X85545.1 - 5 - 6 1176 72

View Full Product Details