Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM240
Entrez Gene ID
339453
Gene Name
transmembrane protein 240
Gene Aliases
C1orf70, SCA21
Chromosome Location
Chr.1: 1534778 - 1540360 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114748.1 NP_001108220.1 3 - 4 389 108
GenBank mRNA BC147018.1 - 4 - 5 418 108
BC147020.1 - 4 - 5 418 108
BU929336.1 - 3 - 4 370 108

View Full Product Details